The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017671.4 | NP_060141.3 | 3 - 4 | 1166 | 107 |
GenBank mRNA | AB105105.1 | - | 3 - 4 | 1166 | 107 |
AF443278.1 | - | 3 - 4 | 735 | 107 | |
AK092195.1 | - | 1 - 2 | 224 | 107 | |
AK314394.1 | - | 3 - 4 | 556 | 107 | |
AY137240.1 | - | 2 - 3 | 377 | 107 | |
BC035882.1 | - | 3 - 4 | 849 | 107 | |
BC040545.1 | - | 3 - 4 | 635 | 107 |