The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129727.2 | NP_001123199.1 | 6 - 7 | 1153 | 81 |
NM_001129728.1 | NP_001123200.1 | 5 - 6 | 1113 | 81 | |
NM_001129729.2 | NP_001123201.1 | 5 - 6 | 1086 | 81 | |
NM_001129731.2 | NP_001123203.1 | 3 - 4 | 1092 | 81 | |
XM_011522985.2 | XP_011521287.1 | 5 - 6 | 1083 | 81 | |
XM_011522986.2 | XP_011521288.1 | 5 - 6 | 1215 | 81 | |
XM_011522987.2 | XP_011521289.1 | 6 - 7 | 1159 | 81 | |
XM_011522988.2 | XP_011521290.1 | 5 - 6 | 1032 | 81 | |
GenBank mRNA | AB197663.1 | - | 4 - 5 | 3349 | 81 |
AB197664.1 | - | 3 - 4 | 3516 | 81 | |
AK024475.1 | - | 5 - 6 | 1048 | 81 | |
AK304723.1 | - | 3 - 4 | 504 | 81 | |
AK308298.1 | - | 5 - 6 | 1022 | 81 | |
BC054486.1 | - | 3 - 4 | 1027 | 81 | |
BC063501.1 | - | 5 - 6 | 1024 | 81 | |
BC082974.1 | - | 6 - 7 | 1121 | 81 | |
BC115390.1 | - | 3 - 4 | 1130 | 81 |