Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNS1
Entrez Gene ID
7145
Gene Name
tensin 1
Gene Aliases
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location
Chr.2: 217799789 - 217978853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308022.1 NP_001294951.1 28 - 29 5103 60
NM_001308023.1 NP_001294952.1 27 - 28 5082 60
NM_022648.5 NP_072174.3 30 - 31 5354 60
XM_011511712.1 XP_011510014.1 28 - 29 5337 60
XM_011511713.1 XP_011510015.1 28 - 29 5322 60
XM_011511715.1 XP_011510017.1 27 - 28 5298 60
XM_011511723.2 XP_011510025.1 28 - 29 5298 60
XM_011511728.1 XP_011510030.1 12 - 13 6390 60
XM_017004801.1 XP_016860290.1 29 - 30 5490 60
XM_017004802.1 XP_016860291.1 28 - 29 5445 60
XM_017004803.1 XP_016860292.1 28 - 29 5406 60
XM_017004804.1 XP_016860293.1 29 - 30 5361 60
XM_017004805.1 XP_016860294.1 28 - 29 5277 60
XM_017004806.1 XP_016860295.1 27 - 28 5250 60
XM_017004807.1 XP_016860296.1 26 - 27 5211 60
XM_017004808.1 XP_016860297.1 25 - 26 4986 60
XM_017004809.1 XP_016860298.1 33 - 34 5834 60
XM_017004810.1 XP_016860299.1 33 - 34 5717 60
XM_017004811.1 XP_016860300.1 31 - 32 5580 60
XM_017004812.1 XP_016860301.1 30 - 31 5530 60
XM_017004813.1 XP_016860302.1 16 - 17 4442 60
XM_017004814.1 XP_016860303.1 16 - 17 4230 60
XM_017004815.1 XP_016860304.1 16 - 17 4296 60
XM_017004816.1 XP_016860305.1 9 - 10 4605 60
XM_017004817.1 XP_016860306.1 14 - 15 6585 60
GenBank mRNA AB209238.1 - 12 - 13 2370 60
AL117537.1 - 6 - 7 562 60
BC014042.2 - 7 - 8 1066 60
BC116187.1 - 27 - 28 5076 60
BC116188.1 - 26 - 27 5052 60
BC126910.1 - 26 - 27 5055 60
BC140942.1 - 30 - 31 5286 60

View Full Product Details