Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TNS1
- Entrez Gene ID
-
7145
- Gene Name
- tensin 1
- Gene Aliases
-
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
- Chromosome Location
-
Chr.2: 217799789 - 217978853 on Build GRCh38
View Full Product Details