Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNS1
Entrez Gene ID
7145
Gene Name
tensin 1
Gene Aliases
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location
Chr.2: 217799789 - 217978853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308022.1 NP_001294951.1 4 - 5 182 63
NM_001308023.1 NP_001294952.1 4 - 5 182 63
NM_022648.5 NP_072174.3 5 - 6 391 63
XM_011511712.1 XP_011510014.1 1 - 2 47 63
XM_011511713.1 XP_011510015.1 1 - 2 47 63
XM_011511715.1 XP_011510017.1 1 - 2 47 63
XM_011511723.2 XP_011510025.1 5 - 6 398 63
XM_017004801.1 XP_016860290.1 1 - 2 47 63
XM_017004802.1 XP_016860291.1 1 - 2 47 63
XM_017004803.1 XP_016860292.1 1 - 2 47 63
XM_017004804.1 XP_016860293.1 1 - 2 47 63
XM_017004805.1 XP_016860294.1 1 - 2 47 63
XM_017004806.1 XP_016860295.1 1 - 2 47 63
XM_017004807.1 XP_016860296.1 1 - 2 47 63
XM_017004808.1 XP_016860297.1 1 - 2 47 63
XM_017004809.1 XP_016860298.1 5 - 6 388 63
XM_017004810.1 XP_016860299.1 5 - 6 271 63
XM_017004811.1 XP_016860300.1 5 - 6 398 63
GenBank mRNA AK001785.2 - 5 - 6 237 63
AK300345.1 - 5 - 6 388 63
AK309785.1 - 7 - 8 491 63
AK310169.1 - 2 - 3 213 63
AL706926.1 - 5 - 6 391 63
BC051304.1 - 2 - 3 258 63
BC071905.1 - 2 - 3 258 63
BC116187.1 - 3 - 4 155 63
BC116188.1 - 3 - 4 155 63
BC126910.1 - 3 - 4 155 63
BC140942.1 - 5 - 6 323 63

View Full Product Details