The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003190.4 | NP_003181.3 | 4 - 5 | 1213 | 73 |
NM_172208.2 | NP_757345.2 | 4 - 5 | 1213 | 73 | |
NM_172209.2 | NP_757346.2 | 3 - 4 | 952 | 73 | |
XM_011514828.2 | XP_011513130.1 | 4 - 5 | 1080 | 73 | |
XM_017011227.1 | XP_016866716.1 | 4 - 5 | 1080 | 73 | |
GenBank mRNA | AB010639.1 | - | 4 - 5 | 880 | 73 |
AF009510.1 | - | 4 - 5 | 883 | 73 | |
AF029750.1 | - | 4 - 5 | 905 | 73 | |
AF314222.1 | - | 4 - 5 | 867 | 73 | |
BC080574.1 | - | 4 - 5 | 1038 | 73 | |
Y13582.1 | - | 4 - 5 | 996 | 73 |