Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIMT1
Entrez Gene ID
27292
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
DIM1, DIMT1L, HSA9761, HUSSY5
Chromosome Location
Chr.5: 62388523 - 62403939 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014473.2 NP_055288.1 1 - 2 207 53
XM_011543323.2 XP_011541625.1 1 - 2 245 53
GenBank mRNA AF102147.1 - 1 - 2 156 53
AK292723.1 - 1 - 2 235 53
AK299480.1 - 1 - 2 197 53
BC002841.2 - 1 - 2 204 53
BC010874.2 - 1 - 2 168 53
BM749030.1 - 1 - 2 217 53

View Full Product Details