Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
5152
Gene Name
phosphodiesterase 9A
Gene Aliases
HSPDE9A2
Chromosome Location
Chr.21: 42653752 - 42775509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001567.1 NP_001001567.1 6 - 7 447 72
NM_001001568.1 NP_001001568.1 4 - 5 298 72
NM_001001569.1 NP_001001569.1 3 - 4 243 72
NM_001001570.1 NP_001001570.1 6 - 7 468 72
NM_001001571.1 NP_001001571.1 5 - 6 376 72
NM_001001572.1 NP_001001572.1 3 - 4 254 72
NM_001001573.1 NP_001001573.1 4 - 5 434 72
NM_001001574.1 NP_001001574.1 5 - 6 369 72
NM_001001575.1 NP_001001575.1 2 - 3 199 72
NM_001001577.1 NP_001001577.1 3 - 4 277 72
NM_001001578.1 NP_001001578.1 4 - 5 321 72
NM_001001579.1 NP_001001579.1 4 - 5 314 72
NM_001001581.1 NP_001001581.1 5 - 6 478 72
NM_001001582.1 NP_001001582.1 6 - 7 556 72
NM_001001583.1 NP_001001583.1 6 - 7 549 72
NM_001001584.2 NP_001001584.1 5 - 6 377 72
NM_001001585.1 NP_001001585.1 5 - 6 501 72
NM_001315533.1 NP_001302462.1 7 - 8 523 72
NM_002606.2 NP_002597.1 7 - 8 627 72
XM_011529598.2 XP_011527900.1 7 - 8 549 72
XM_011529600.2 XP_011527902.1 7 - 8 600 72
XM_017028366.1 XP_016883855.1 5 - 6 365 72
XM_017028367.1 XP_016883856.1 7 - 8 600 72
GenBank mRNA AF048837.1 - 7 - 8 624 72
AF067223.1 - 7 - 8 576 72
AF067224.1 - 6 - 7 395 72
AF067225.1 - 4 - 5 237 72
AF067226.1 - 3 - 4 182 72
AK001165.1 - 4 - 5 229 72
AK001307.1 - 6 - 7 418 72
AK294112.1 - 4 - 5 264 72
AK314679.1 - 7 - 8 600 72
AY196299.1 - 6 - 7 441 72
AY196300.1 - 5 - 6 349 72
AY196301.1 - 3 - 4 227 72
AY196302.1 - 4 - 5 407 72
AY196303.1 - 5 - 6 342 72
AY196304.1 - 2 - 3 172 72
AY196306.1 - 3 - 4 250 72
AY196307.1 - 4 - 5 294 72
AY196308.1 - 4 - 5 287 72
AY196310.1 - 5 - 6 451 72
AY196311.1 - 6 - 7 529 72
AY196312.1 - 6 - 7 522 72
AY196313.1 - 5 - 6 349 72
AY196314.1 - 5 - 6 474 72
AY242121.1 - 5 - 6 313 72
AY701187.1 - 7 - 8 520 72
BC009047.1 - 6 - 7 421 72
BT007016.1 - 6 - 7 384 72
KU178246.1 - 6 - 7 384 72
KU178247.1 - 7 - 8 564 72
KU178248.1 - 3 - 4 180 72

View Full Product Details