Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
Chromosome Location
Chr.6: 36196773 - 36232790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015695.2 NP_056510.2 12 - 13 3662 75
XM_005249010.2 XP_005249067.1 12 - 13 3544 75
XM_005249011.3 XP_005249068.1 12 - 13 3668 75
XM_011514489.1 XP_011512791.1 12 - 13 3566 75
XM_017010743.1 XP_016866232.1 9 - 10 1780 75
GenBank mRNA AB033112.1 - 12 - 13 3662 75
AK025134.1 - 5 - 6 722 75
AK074362.1 - 9 - 10 1800 75
AK172787.1 - 3 - 4 300 75
BC117387.1 - 9 - 10 2659 75
BC143917.1 - 9 - 10 2659 75
BC143918.1 - 7 - 8 2467 75

View Full Product Details