The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317214.1 | NP_001304143.1 | 6 - 7 | 1676 | 77 |
NM_004361.3 | NP_004352.2 | 6 - 7 | 1676 | 77 | |
NM_033646.2 | NP_387450.1 | 6 - 7 | 1306 | 77 | |
XM_017025523.1 | XP_016881012.1 | 6 - 7 | 1992 | 77 | |
GenBank mRNA | AB035301.1 | - | 6 - 7 | 1407 | 77 |
AJ007611.1 | - | 6 - 7 | 1304 | 77 | |
BC036786.1 | - | 6 - 7 | 1676 | 77 |