Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDC1
Entrez Gene ID
55706
Gene Name
NDC1 transmembrane nucleoporin
Gene Aliases
NET3, TMEM48
Chromosome Location
Chr.1: 53765460 - 53838552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL354612.1 - 13 - 14 1701 149

View Full Product Details