The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168551.1 | NP_001162023.1 | 6 - 7 | 875 | 120 |
NM_018087.4 | NP_060557.3 | 6 - 7 | 995 | 120 | |
NR_033142.1 | 5 - 6 | 874 | 120 | ||
XM_011541766.2 | XP_011540068.1 | 6 - 7 | 837 | 120 | |
GenBank mRNA | AK001269.1 | - | 6 - 7 | 878 | 120 |
AK295000.1 | - | 6 - 7 | 659 | 120 | |
AK298909.1 | - | 5 - 6 | 618 | 120 | |
AL354612.1 | - | 6 - 7 | 878 | 120 | |
AL354613.1 | - | 6 - 7 | 875 | 120 | |
BC003082.2 | - | 6 - 7 | 945 | 120 | |
BX648031.1 | - | 5 - 6 | 605 | 120 | |
DQ141696.1 | - | 6 - 7 | 704 | 120 |