The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014698.2 | NP_055513.2 | 20 - 21 | 2530 | 64 |
XM_006711841.3 | XP_006711904.1 | 18 - 19 | 2023 | 64 | |
XM_011544328.2 | XP_011542630.1 | 20 - 21 | 2382 | 64 | |
XM_011544329.2 | XP_011542631.1 | 20 - 21 | 3649 | 64 | |
XM_011544330.2 | XP_011542632.1 | 19 - 20 | 2339 | 64 | |
XM_011544331.2 | XP_011542633.1 | 19 - 20 | 2126 | 64 | |
XM_011544332.2 | XP_011542634.1 | 19 - 20 | 2161 | 64 | |
XM_017002916.1 | XP_016858405.1 | 19 - 20 | 2613 | 64 | |
GenBank mRNA | AB018335.1 | - | 20 - 21 | 2158 | 64 |
AK222934.1 | - | 20 - 21 | 2160 | 64 | |
AK222944.1 | - | 20 - 21 | 2160 | 64 | |
BC030245.1 | - | 20 - 21 | 2163 | 64 |