The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015470.2 | NP_056285.1 | 3 - 4 | 1812 | 95 |
GenBank mRNA | AB020664.1 | - | 3 - 4 | 1812 | 95 |
AF153085.1 | - | 3 - 4 | 1260 | 95 | |
AF334812.1 | - | 3 - 4 | 1811 | 95 | |
AK023823.1 | - | 2 - 3 | 1477 | 95 | |
BC035013.1 | - | 3 - 4 | 1735 | 95 |