The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130079.2 | NP_001123551.1 | 5 - 6 | 1055 | 67 |
NM_001286072.1 | NP_001273001.1 | 6 - 7 | 2079 | 67 | |
NM_001321324.1 | NP_001308253.1 | 5 - 6 | 956 | 67 | |
NM_020963.4 | NP_066014.1 | 5 - 6 | 1331 | 67 | |
XM_005270869.4 | XP_005270926.1 | 6 - 7 | 1282 | 67 | |
XM_005270870.4 | XP_005270927.1 | 6 - 7 | 1298 | 67 | |
XM_017001318.1 | XP_016856807.1 | 6 - 7 | 1889 | 67 | |
XM_017001319.1 | XP_016856808.1 | 5 - 6 | 956 | 67 | |
GenBank mRNA | AB046851.1 | - | 6 - 7 | 1693 | 67 |
AK023297.1 | - | 5 - 6 | 933 | 67 | |
AK074174.1 | - | 4 - 5 | 907 | 67 | |
AK226011.1 | - | 5 - 6 | 958 | 67 | |
AK301229.1 | - | 5 - 6 | 684 | 67 | |
AK308874.1 | - | 5 - 6 | 967 | 67 | |
AK310512.1 | - | 5 - 6 | 1130 | 67 | |
AL833353.1 | - | 5 - 6 | 1222 | 67 | |
BC002548.1 | - | 4 - 5 | 902 | 67 | |
BC009312.2 | - | 4 - 5 | 926 | 67 | |
BC025339.1 | - | 4 - 5 | 926 | 67 | |
BX647787.1 | - | 6 - 7 | 1671 | 67 |