The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002754.4 | NP_002745.1 | 8 - 9 | 786 | 78 |
GenBank mRNA | AB451221.1 | - | 9 - 10 | 687 | 78 |
AB451344.1 | - | 9 - 10 | 687 | 78 | |
AF004709.1 | - | 8 - 9 | 735 | 78 | |
AF015256.1 | - | 8 - 9 | 747 | 78 | |
AF092535.1 | - | 8 - 9 | 708 | 78 | |
AF100546.1 | - | 8 - 9 | 689 | 78 | |
AK308018.1 | - | 8 - 9 | 767 | 78 | |
BC000433.2 | - | 8 - 9 | 728 | 78 | |
BC001641.2 | - | 8 - 9 | 728 | 78 | |
BC004428.1 | - | 8 - 9 | 728 | 78 | |
BM921729.1 | - | 8 - 9 | 79 | ||
BT007221.1 | - | 8 - 9 | 687 | 78 | |
CR536490.1 | - | 8 - 9 | 687 | 78 | |
CR541687.1 | - | 8 - 9 | 687 | 78 | |
KU178281.1 | - | 8 - 9 | 687 | 78 | |
KU178282.1 | - | 8 - 9 | 720 | 78 | |
U93232.1 | - | 8 - 9 | 694 | 78 | |
Y10488.1 | - | 8 - 9 | 709 | 78 |