Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM7
Entrez Gene ID
54822
Gene Name
transient receptor potential cation channel subfamily M member 7
Gene Aliases
ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK
Chromosome Location
Chr.15: 50557158 - 50686835 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301212.1 NP_001288141.1 6 - 7 947 126
NM_017672.5 NP_060142.3 6 - 7 947 126
XM_005254486.3 XP_005254543.1 6 - 7 942 126
XM_017022350.1 XP_016877839.1 6 - 7 707 126
XM_017022351.1 XP_016877840.1 6 - 7 707 126
XM_017022352.1 XP_016877841.1 6 - 7 707 126
XM_017022353.1 XP_016877842.1 6 - 7 707 126
XM_017022354.1 XP_016877843.1 6 - 7 772 126
XM_017022355.1 XP_016877844.1 4 - 5 574 126
XM_017022356.1 XP_016877845.1 6 - 7 707 126
GenBank mRNA AF346629.2 - 6 - 7 903 126
AK000124.1 - 6 - 7 878 126
AY032950.1 - 6 - 7 936 126
BC128987.1 - 6 - 7 758 126
BC128988.1 - 6 - 7 758 126

View Full Product Details