The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301212.1 | NP_001288141.1 | 24 - 25 | 3751 | 163 |
NM_017672.5 | NP_060142.3 | 24 - 25 | 3751 | 163 | |
XM_005254486.3 | XP_005254543.1 | 24 - 25 | 3746 | 163 | |
XM_017022350.1 | XP_016877839.1 | 24 - 25 | 3511 | 163 | |
XM_017022351.1 | XP_016877840.1 | 24 - 25 | 3511 | 163 | |
XM_017022352.1 | XP_016877841.1 | 24 - 25 | 3511 | 163 | |
XM_017022353.1 | XP_016877842.1 | 24 - 25 | 3511 | 163 | |
XM_017022354.1 | XP_016877843.1 | 24 - 25 | 3576 | 163 | |
XM_017022355.1 | XP_016877844.1 | 22 - 23 | 3378 | 163 | |
GenBank mRNA | AF346629.2 | - | 24 - 25 | 3707 | 163 |
AK172800.1 | - | 1 - 2 | 196 | 163 | |
AY032950.1 | - | 24 - 25 | 3740 | 163 |