Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRPM7
Entrez Gene ID
54822
Gene Name
transient receptor potential cation channel subfamily M member 7
Gene Aliases
ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK
Chromosome Location
Chr.15: 50557158 - 50686835 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301212.1 NP_001288141.1 32 - 33 4946 102
NM_017672.5 NP_060142.3 32 - 33 4949 102
XM_005254486.3 XP_005254543.1 33 - 34 5004 102
XM_017022350.1 XP_016877839.1 33 - 34 4769 102
XM_017022351.1 XP_016877840.1 33 - 34 4766 102
XM_017022352.1 XP_016877841.1 32 - 33 4709 102
XM_017022353.1 XP_016877842.1 32 - 33 4706 102
XM_017022354.1 XP_016877843.1 33 - 34 4834 102
XM_017022355.1 XP_016877844.1 31 - 32 4636 102
GenBank mRNA AF346629.2 - 32 - 33 4902 102
AK172800.1 - 9 - 10 1394 102
AY032950.1 - 32 - 33 4938 102

View Full Product Details