Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRPM7
Entrez Gene ID
54822
Gene Name
transient receptor potential cation channel subfamily M member 7
Gene Aliases
ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK
Chromosome Location
Chr.15: 50557158 - 50686835 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301212.1 NP_001288141.1 35 - 36 5377 100
NM_017672.5 NP_060142.3 35 - 36 5380 100
XM_005254486.3 XP_005254543.1 36 - 37 5435 100
XM_017022350.1 XP_016877839.1 36 - 37 5200 100
XM_017022351.1 XP_016877840.1 36 - 37 5197 100
XM_017022352.1 XP_016877841.1 35 - 36 5140 100
XM_017022353.1 XP_016877842.1 35 - 36 5137 100
XM_017022354.1 XP_016877843.1 36 - 37 5265 100
XM_017022355.1 XP_016877844.1 34 - 35 5067 100
GenBank mRNA AF346629.2 - 35 - 36 5333 100
AK075193.1 - 1 - 2 85 100
AK098584.1 - 6 - 7 655 100
AK172800.1 - 12 - 13 1825 100
AY032950.1 - 35 - 36 5369 100
BC128987.1 - 22 - 23 3014 100
BC128988.1 - 22 - 23 3014 100

View Full Product Details