Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS5
Entrez Gene ID
64969
Gene Name
mitochondrial ribosomal protein S5
Gene Aliases
MRP-S5, S5mt
Chromosome Location
Chr.2: 95085391 - 95122027 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321995.1 NP_001308924.1 11 - 12 1124 67
NM_001321996.1 NP_001308925.1 11 - 12 1224 67
NM_001321997.1 NP_001308926.1 11 - 12 1045 67
NM_031902.4 NP_114108.1 11 - 12 1303 67
NR_136152.1 10 - 11 1133 67
NR_136153.1 10 - 11 1212 67
GenBank mRNA AB049940.1 - 11 - 12 1184 67
AK055390.1 - 1 - 2 1917 67
AK058160.1 - 10 - 11 986 67
AK124892.1 - 1 - 2 3305 67
BC000219.2 - 7 - 8 618 67
BC014172.2 - 11 - 12 1276 67

View Full Product Details