The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003593.2 | NP_003584.2 | 5 - 6 | 959 | 87 |
XM_005258046.3 | XP_005258103.1 | 6 - 7 | 1157 | 87 | |
XM_011525358.1 | XP_011523660.1 | 6 - 7 | 1160 | 87 | |
XM_011525359.1 | XP_011523661.1 | 6 - 7 | 1143 | 87 | |
XM_011525362.1 | XP_011523664.1 | 6 - 7 | 1041 | 87 | |
XM_011525367.1 | XP_011523669.1 | 4 - 5 | 948 | 87 | |
XM_011525368.2 | XP_011523670.1 | 4 - 5 | 577 | 87 | |
XM_011525369.2 | XP_011523671.1 | 4 - 5 | 725 | 87 | |
XM_011525370.2 | XP_011523672.1 | 4 - 5 | 722 | 87 | |
XM_017025228.1 | XP_016880717.1 | 6 - 7 | 1172 | 87 | |
XM_017025229.1 | XP_016880718.1 | 6 - 7 | 1143 | 87 | |
XM_017025230.1 | XP_016880719.1 | 6 - 7 | 1143 | 87 | |
XM_017025231.1 | XP_016880720.1 | 6 - 7 | 1143 | 87 | |
GenBank mRNA | AK313878.1 | - | 6 - 7 | 1128 | 87 |
Y11739.1 | - | 5 - 6 | 959 | 87 |