The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206480.2 | NP_001193409.1 | 16 - 17 | 1981 | 64 |
NM_001206482.1 | NP_001193411.1 | 16 - 17 | 2151 | 64 | |
NM_014800.10 | NP_055615.8 | 16 - 17 | 1792 | 64 | |
XM_005249919.2 | XP_005249976.1 | 17 - 18 | 2345 | 64 | |
XM_006715805.1 | XP_006715868.1 | 16 - 17 | 1637 | 64 | |
XM_011515654.1 | XP_011513956.1 | 17 - 18 | 3030 | 64 | |
XM_017012838.1 | XP_016868327.1 | 17 - 18 | 3215 | 64 | |
XM_017012839.1 | XP_016868328.1 | 16 - 17 | 1579 | 64 | |
GenBank mRNA | AF398885.1 | - | 15 - 16 | 1443 | 64 |
AK126565.1 | - | 4 - 5 | 832 | 64 | |
AK291784.1 | - | 16 - 17 | 2151 | 64 | |
AK296253.1 | - | 13 - 14 | 1312 | 64 | |
AK314349.1 | - | 16 - 17 | 1581 | 64 | |
AL136787.1 | - | 16 - 17 | 1693 | 64 | |
BC114341.2 | - | 16 - 17 | 1530 | 64 |