Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN31
Entrez Gene ID
6302
Gene Name
tetraspanin 31
Gene Aliases
SAS
Chromosome Location
Chr.12: 57744090 - 57748243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005981.3 NP_005972.1 4 - 5 595 112
XM_005269074.3 XP_005269131.2 4 - 5 1520 112
XM_017019793.1 XP_016875282.1 2 - 3 324 112
GenBank mRNA AK090502.1 - 2 - 3 998 112
AK294128.1 - 4 - 5 548 112
AK315765.1 - 4 - 5 716 112
BC010377.1 - 4 - 5 548 112
CR407647.1 - 4 - 5 441 112
U01160.1 - 4 - 5 543 112

View Full Product Details