Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
Chromosome Location
Chr.7: 95772506 - 96110322 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135556.1 NP_001129028.1 7 - 8 862 149
NM_001135557.1 NP_001129029.1 6 - 7 802 149
NM_001278421.1 NP_001265350.1 6 - 7 853 149
NM_001278422.1 NP_001265351.1 6 - 7 802 149
NM_004411.4 NP_004402.1 7 - 8 913 149
XM_011515861.1 XP_011514163.1 7 - 8 760 149
XM_011515862.1 XP_011514164.1 7 - 8 709 149
XM_011515863.2 XP_011514165.1 7 - 8 867 149
XM_017011804.1 XP_016867293.1 7 - 8 1500 149
XM_017011805.1 XP_016867294.1 7 - 8 687 149
XM_017011806.1 XP_016867295.1 7 - 8 862 149
XM_017011807.1 XP_016867296.1 7 - 8 867 149
GenBank mRNA AF063228.1 - 7 - 8 823 149
AF123074.1 - 6 - 7 711 149
AK091339.1 - 8 - 9 762 149
AK297427.1 - 6 - 7 714 149
AK309419.1 - 7 - 8 685 149
AK313148.1 - 7 - 8 860 149
BC022540.1 - 6 - 7 762 149
BC032945.2 - 7 - 8 814 149
BC037854.1 - 7 - 8 804 149
BC053542.1 - 7 - 8 802 149

View Full Product Details