The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012294.3 | NP_036426.3 | 15 - 16 | 1409 | 95 |
XM_005249914.2 | XP_005249971.1 | 15 - 16 | 1628 | 95 | |
XM_005249916.1 | XP_005249973.1 | 15 - 16 | 1572 | 95 | |
XM_011515652.2 | XP_011513954.1 | 7 - 8 | 3382 | 95 | |
XM_017012837.1 | XP_016868326.1 | 13 - 14 | 1313 | 95 | |
GenBank mRNA | AK295859.1 | - | 5 - 6 | 894 | 95 |
BC039203.1 | - | 5 - 6 | 720 | 95 | |
BP349215.1 | - | 6 - 7 | 467 | 95 | |
D87467.1 | - | 5 - 6 | 701 | 95 | |
KU178490.1 | - | 5 - 6 | 646 | 95 | |
KU178491.1 | - | 5 - 6 | 646 | 95 |