Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBP
Entrez Gene ID
6908
Gene Name
TATA-box binding protein
Gene Aliases
GTF2D, GTF2D1, HDL4, SCA17, TFIID
Chromosome Location
Chr.6: 170554333 - 170572870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC109054.1 - 1 - 2 53 82
M55654.1 - 1 - 2 82

View Full Product Details