The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130172.1 | NP_001123644.1 | 1 - 2 | 220 | 59 |
NM_001130173.1 | NP_001123645.1 | 1 - 2 | 220 | 59 | |
NM_001161656.1 | NP_001155128.1 | 1 - 2 | 220 | 59 | |
NM_001161657.1 | NP_001155129.1 | 1 - 2 | 220 | 59 | |
NM_001161658.1 | NP_001155130.1 | 1 - 2 | 220 | 59 | |
NM_001161659.1 | NP_001155131.1 | 1 - 2 | 220 | 59 | |
NM_001161660.1 | NP_001155132.1 | 1 - 2 | 220 | 59 | |
NM_005375.3 | NP_005366.2 | 1 - 2 | 220 | 59 | |
NR_134958.1 | 1 - 2 | 220 | 59 | ||
NR_134959.1 | 1 - 2 | 220 | 59 | ||
NR_134960.1 | 1 - 2 | 220 | 59 | ||
NR_134961.1 | 1 - 2 | 220 | 59 | ||
NR_134962.1 | 1 - 2 | 220 | 59 | ||
NR_134963.1 | 1 - 2 | 220 | 59 | ||
NR_134964.1 | 1 - 2 | 220 | 59 | ||
NR_134965.1 | 1 - 2 | 220 | 59 | ||
GenBank mRNA | AF104863.1 | - | 1 - 2 | 98 | 59 |
AJ606317.1 | - | 1 - 2 | 220 | 59 | |
AJ606318.1 | - | 1 - 2 | 220 | 59 | |
AJ606319.1 | - | 1 - 2 | 220 | 59 | |
AJ606320.1 | - | 1 - 2 | 220 | 59 | |
AJ606321.1 | - | 1 - 2 | 220 | 59 | |
AJ606322.1 | - | 1 - 2 | 220 | 59 | |
AJ606323.1 | - | 1 - 2 | 220 | 59 | |
AJ606324.1 | - | 1 - 2 | 220 | 59 | |
AJ616235.1 | - | 1 - 2 | 135 | 59 | |
BC064955.1 | - | 1 - 2 | 172 | 59 | |
M15024.1 | - | 1 - 2 | 134 | 59 | |
X52125.1 | - | 1 - 2 | 218 | 59 |