The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281748.1 | NP_001268677.1 | 17 - 18 | 2203 | 65 |
NM_001281749.1 | NP_001268678.1 | 16 - 17 | 2079 | 65 | |
NM_025164.4 | NP_079440.3 | 16 - 17 | 2079 | 65 | |
XM_005271481.2 | XP_005271538.1 | 17 - 18 | 2261 | 65 | |
XM_005271482.4 | XP_005271539.2 | 16 - 17 | 2117 | 65 | |
XM_005271484.3 | XP_005271541.2 | 16 - 17 | 2117 | 65 | |
XM_005271485.3 | XP_005271542.1 | 4 - 5 | 599 | 65 | |
XM_011542721.2 | XP_011541023.1 | 17 - 18 | 2223 | 65 | |
XM_011542722.1 | XP_011541024.1 | 17 - 18 | 2261 | 65 | |
XM_011542723.2 | XP_011541025.1 | 17 - 18 | 2104 | 65 | |
XM_011542724.2 | XP_011541026.1 | 16 - 17 | 1958 | 65 | |
XM_011542725.2 | XP_011541027.1 | 15 - 16 | 1841 | 65 | |
XM_017017424.1 | XP_016872913.1 | 15 - 16 | 1973 | 65 | |
XM_017017425.1 | XP_016872914.1 | 17 - 18 | 2299 | 65 | |
XM_017017426.1 | XP_016872915.1 | 15 - 16 | 2173 | 65 | |
XM_017017427.1 | XP_016872916.1 | 12 - 13 | 1576 | 65 | |
GenBank mRNA | AB209327.1 | - | 9 - 10 | 1089 | 65 |
AK122838.1 | - | 2 - 3 | 1521 | 65 | |
BC063887.1 | - | 3 - 4 | 382 | 65 | |
BC128510.1 | - | 16 - 17 | 1941 | 65 | |
BC128511.1 | - | 17 - 18 | 2059 | 65 |