The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281748.1 | NP_001268677.1 | 7 - 8 | 995 | 86 |
NM_001281749.1 | NP_001268678.1 | 6 - 7 | 871 | 86 | |
NM_025164.4 | NP_079440.3 | 6 - 7 | 871 | 86 | |
XM_005271481.2 | XP_005271538.1 | 6 - 7 | 909 | 86 | |
XM_005271482.4 | XP_005271539.2 | 6 - 7 | 909 | 86 | |
XM_005271484.3 | XP_005271541.2 | 6 - 7 | 909 | 86 | |
XM_011542721.2 | XP_011541023.1 | 6 - 7 | 871 | 86 | |
XM_011542722.1 | XP_011541024.1 | 6 - 7 | 909 | 86 | |
XM_011542723.2 | XP_011541025.1 | 6 - 7 | 752 | 86 | |
XM_011542724.2 | XP_011541026.1 | 5 - 6 | 606 | 86 | |
XM_011542725.2 | XP_011541027.1 | 4 - 5 | 489 | 86 | |
XM_011542726.1 | XP_011541028.1 | 6 - 7 | 909 | 86 | |
XM_017017424.1 | XP_016872913.1 | 6 - 7 | 909 | 86 | |
XM_017017425.1 | XP_016872914.1 | 6 - 7 | 947 | 86 | |
XM_017017426.1 | XP_016872915.1 | 4 - 5 | 821 | 86 | |
XM_017017427.1 | XP_016872916.1 | 2 - 3 | 178 | 86 | |
GenBank mRNA | BC128510.1 | - | 6 - 7 | 733 | 86 |
BC128511.1 | - | 7 - 8 | 851 | 86 |