Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location
Chr.19: 49690672 - 49713731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136052.2 NP_001129524.1 6 - 7 869 88
NM_001199752.2 NP_001186681.1 6 - 7 933 88
NM_001199753.1 NP_001186682.1 6 - 7 869 88
NM_152359.2 NP_689572.1 5 - 6 800 88
NR_108072.1 7 - 8 1031 88
XM_005258505.2 XP_005258562.1 6 - 7 906 88
XM_005258506.4 XP_005258563.1 5 - 6 822 88
XM_006723009.3 XP_006723072.1 4 - 5 539 88
XM_011526438.2 XP_011524740.1 6 - 7 962 88
XM_011526439.2 XP_011524741.1 6 - 7 1026 88
XM_011526440.2 XP_011524742.1 6 - 7 878 88
XM_017026265.1 XP_016881754.1 6 - 7 925 88
XM_017026266.1 XP_016881755.1 6 - 7 879 88
XM_017026267.1 XP_016881756.1 5 - 6 696 88
XM_017026268.1 XP_016881757.1 4 - 5 539 88
XM_017026269.1 XP_016881758.1 4 - 5 649 88
XM_017026270.1 XP_016881759.1 4 - 5 417 88
XM_017026271.1 XP_016881760.1 4 - 5 539 88
XM_017026272.1 XP_016881761.1 5 - 6 733 88
XM_017026273.1 XP_016881762.1 5 - 6 718 88
GenBank mRNA AF357970.1 - 5 - 6 758 88
AK096516.1 - 7 - 8 1027 88
AK289713.1 - 6 - 7 925 88
AK290092.1 - 6 - 7 841 88
AK299866.1 - 4 - 5 528 88
BC029104.1 - 6 - 7 833 88

View Full Product Details