The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042625.1 | NP_001036090.1 | 2 - 3 | 230 | 129 |
NM_144647.3 | NP_653248.3 | 2 - 3 | 262 | 129 | |
XM_006714444.3 | XP_006714507.1 | 2 - 3 | 212 | 129 | |
XM_006714445.3 | XP_006714508.1 | 2 - 3 | 220 | 129 | |
GenBank mRNA | BC017586.2 | - | 2 - 3 | 264 | 129 |
CD687387.1 | - | 2 - 3 | 222 | 129 |