The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042625.1 | NP_001036090.1 | 4 - 5 | 616 | 115 |
NM_144647.3 | NP_653248.3 | 4 - 5 | 648 | 115 | |
XM_006714444.3 | XP_006714507.1 | 4 - 5 | 598 | 115 | |
GenBank mRNA | BC017586.2 | - | 4 - 5 | 650 | 115 |
BM975803.1 | - | 1 - 2 | 115 |