Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNOT4
Entrez Gene ID
4850
Gene Name
CCR4-NOT transcription complex subunit 4
Gene Aliases
CLONE243, NOT4, NOT4H
Chromosome Location
Chr.7: 135361795 - 135510127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008225.2 NP_001008226.1 6 - 7 1017 117
NM_001190847.1 NP_001177776.1 6 - 7 1017 117
NM_001190848.1 NP_001177777.1 6 - 7 1017 117
NM_001190849.1 NP_001177778.1 6 - 7 1017 117
NM_001190850.1 NP_001177779.1 6 - 7 1017 117
NM_013316.3 NP_037448.2 6 - 7 1017 117
XM_017012235.1 XP_016867724.1 6 - 7 878 117
GenBank mRNA AF091094.1 - 3 - 4 281 117
AF180475.1 - 5 - 6 686 117
AK074671.1 - 3 - 4 244 117
AK300365.1 - 6 - 7 983 117
AK314540.1 - 6 - 7 982 117
AL389980.1 - 6 - 7 971 117
BC035590.1 - 6 - 7 977 117
U71267.1 - 6 - 7 967 117
U71268.1 - 6 - 7 967 117

View Full Product Details