The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004342.6 | NP_004333.1 | 1 - 2 | 332 | 126 |
NM_033138.3 | NP_149129.2 | 1 - 2 | 332 | 126 | |
NM_033157.3 | NP_149347.2 | 1 - 2 | 332 | 126 | |
GenBank mRNA | AF247820.3 | - | 1 - 2 | 95 | 126 |
AK289686.1 | - | 1 - 2 | 325 | 126 | |
AK298524.1 | - | 1 - 2 | 111 | 126 | |
AK304729.1 | - | 1 - 2 | 108 | 126 | |
BC040354.1 | - | 1 - 2 | 67 | 126 | |
CD359921.1 | - | 1 - 2 | 310 | 126 | |
DB090561.1 | - | 1 - 2 | 332 | 126 | |
M83216.1 | - | 1 - 2 | 95 | 126 |