The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004629.1 | NP_004620.1 | 3 - 4 | 792 | 65 |
GenBank mRNA | AJ007669.1 | - | 3 - 4 | 792 | 65 |
AK293427.1 | - | 4 - 5 | 482 | 65 | |
AK312987.1 | - | 3 - 4 | 717 | 65 | |
BC000032.2 | - | 3 - 4 | 438 | 65 | |
BC011623.2 | - | 3 - 4 | 565 | 65 | |
BT009813.1 | - | 3 - 4 | 300 | 65 | |
KU159422.1 | - | 3 - 4 | 823 | 65 | |
KU178053.1 | - | 3 - 4 | 300 | 65 | |
KU178054.1 | - | 3 - 4 | 300 | 65 | |
KU178055.1 | - | 3 - 4 | 300 | 65 | |
U70310.1 | - | 3 - 4 | 434 | 65 |