The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135999.1 | NP_001129471.1 | 5 - 6 | 3551 | 62 |
NM_014806.3 | NP_055621.1 | 5 - 6 | 3295 | 62 | |
NR_052015.2 | 5 - 6 | 2970 | 62 | ||
XM_006716896.2 | XP_006716959.1 | 4 - 5 | 3142 | 62 | |
GenBank mRNA | AB002373.2 | - | 5 - 6 | 3137 | 62 |
BC064843.1 | - | 4 - 5 | 2339 | 62 | |
BC132766.1 | - | 4 - 5 | 3080 | 62 | |
BC132770.1 | - | 4 - 5 | 3080 | 62 | |
BC146654.1 | - | 5 - 6 | 3269 | 62 | |
BC150211.1 | - | 5 - 6 | 2882 | 62 | |
BC150262.1 | - | 5 - 6 | 3148 | 62 |