The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000222.2 | NP_000213.1 | 6 - 7 | 1203 | 127 |
NM_001093772.1 | NP_001087241.1 | 6 - 7 | 1203 | 127 | |
XM_005265740.1 | XP_005265797.1 | 6 - 7 | 1177 | 127 | |
XM_005265741.1 | XP_005265798.1 | 6 - 7 | 1177 | 127 | |
XM_005265742.2 | XP_005265799.1 | 6 - 7 | 1177 | 127 | |
XM_017008178.1 | XP_016863667.1 | 6 - 7 | 1174 | 127 | |
XM_017008179.1 | XP_016863668.1 | 6 - 7 | 1177 | 127 | |
XM_017008180.1 | XP_016863669.1 | 6 - 7 | 1174 | 127 | |
GenBank mRNA | AK304031.1 | - | 6 - 7 | 1174 | 127 |
BC071593.1 | - | 6 - 7 | 1192 | 127 | |
EU826594.1 | - | 6 - 7 | 1116 | 127 | |
GU983671.1 | - | 6 - 7 | 1121 | 127 | |
HM015525.1 | - | 6 - 7 | 1121 | 127 | |
HM015526.1 | - | 6 - 7 | 1121 | 127 | |
KT326922.1 | - | 6 - 7 | 1134 | 127 | |
X06182.1 | - | 6 - 7 | 1137 | 127 |