The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032773.3 | NP_116162.1 | 6 - 7 | 947 | 61 |
XM_005269362.2 | XP_005269419.1 | 6 - 7 | 1318 | 61 | |
XM_005269363.2 | XP_005269420.1 | 6 - 7 | 1318 | 61 | |
XM_005269365.2 | XP_005269422.1 | 6 - 7 | 1318 | 61 | |
XM_005269367.2 | XP_005269424.1 | 6 - 7 | 1318 | 61 | |
XM_006713791.3 | XP_006713854.1 | 6 - 7 | 1318 | 61 | |
XM_006713792.3 | XP_006713855.1 | 6 - 7 | 1318 | 61 | |
XM_006713793.2 | XP_006713856.1 | 6 - 7 | 1318 | 61 | |
XM_011513242.2 | XP_011511544.1 | 6 - 7 | 1318 | 61 | |
XM_011513243.2 | XP_011511545.1 | 6 - 7 | 1318 | 61 | |
XM_017007350.1 | XP_016862839.1 | 6 - 7 | 1318 | 61 | |
XM_017007351.1 | XP_016862840.1 | 6 - 7 | 1318 | 61 | |
XM_017007352.1 | XP_016862841.1 | 6 - 7 | 1318 | 61 | |
GenBank mRNA | AK055819.1 | - | 1 - 2 | 53 | 61 |
AK293745.1 | - | 6 - 7 | 936 | 61 | |
AK300466.1 | - | 3 - 4 | 531 | 61 | |
AK303521.1 | - | 6 - 7 | 909 | 61 | |
BC007504.2 | - | 6 - 7 | 899 | 61 |