Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP5
Entrez Gene ID
6425
Gene Name
secreted frizzled related protein 5
Gene Aliases
SARP3
Chromosome Location
Chr.10: 97766751 - 97771999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003015.3 NP_003006.2 2 - 3 775 103
GenBank mRNA AF017988.1 - 2 - 3 824 103
AF117758.1 - 2 - 3 756 103
BC050435.1 - 2 - 3 732 103

View Full Product Details