Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PMS1
- Entrez Gene ID
-
5378
- Gene Name
- PMS1 homolog 1, mismatch repair system component
- Gene Aliases
-
HNPCC3, MLH2, PMSL1, hPMS1
- Chromosome Location
-
Chr.2: 189784085 - 189877629 on Build GRCh38
View Full Product Details