The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015542.3 | NP_056357.1 | 13 - 14 | 2675 | 79 |
NM_080599.2 | NP_542166.1 | 13 - 14 | 2845 | 79 | |
XM_017016034.1 | XP_016871523.1 | 12 - 13 | 2331 | 79 | |
GenBank mRNA | AF301013.1 | - | 13 - 14 | 2668 | 79 |
AF318574.1 | - | 13 - 14 | 2645 | 79 | |
AK310925.1 | - | 13 - 14 | 2634 | 79 | |
AL080198.1 | - | 7 - 8 | 845 | 79 | |
BC114964.1 | - | 13 - 14 | 2639 | 79 | |
BC115737.1 | - | 13 - 14 | 2639 | 79 |