The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015542.3 | NP_056357.1 | 7 - 8 | 1869 | 70 |
NM_080599.2 | NP_542166.1 | 7 - 8 | 2039 | 70 | |
XM_011519449.2 | XP_011517751.1 | 7 - 8 | 1917 | 70 | |
XM_017016034.1 | XP_016871523.1 | 6 - 7 | 1525 | 70 | |
GenBank mRNA | AB037829.1 | - | 6 - 7 | 2238 | 70 |
AF301013.1 | - | 7 - 8 | 1862 | 70 | |
AF318574.1 | - | 7 - 8 | 1839 | 70 | |
AK310925.1 | - | 7 - 8 | 1828 | 70 | |
AL080198.1 | - | 1 - 2 | 39 | 70 | |
AY013249.1 | - | 6 - 7 | 1788 | 70 | |
BC114964.1 | - | 7 - 8 | 1833 | 70 | |
BC115737.1 | - | 7 - 8 | 1833 | 70 |