The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080491.3 | NP_001073960.1 | 11 - 12 | 925 | 151 |
NM_014688.3 | NP_055503.1 | 12 - 13 | 1256 | 151 | |
XM_006717542.3 | XP_006717605.1 | 13 - 14 | 1082 | 151 | |
XM_011519762.2 | XP_011518064.1 | 14 - 15 | 1415 | 151 | |
XM_011519763.2 | XP_011518065.1 | 7 - 8 | 1416 | 151 | |
XM_017016971.1 | XP_016872460.1 | 13 - 14 | 1089 | 151 | |
XM_017016972.1 | XP_016872461.1 | 13 - 14 | 1227 | 151 | |
XM_017016973.1 | XP_016872462.1 | 11 - 12 | 4252 | 151 | |
GenBank mRNA | AB449916.1 | - | 10 - 11 | 826 | 151 |
AK292944.1 | - | 12 - 13 | 1046 | 151 | |
BC042943.2 | - | 12 - 13 | 1094 | 151 | |
D13644.1 | - | 12 - 13 | 1105 | 151 |