The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080491.3 | NP_001073960.1 | 3 - 4 | 256 | 94 |
NM_014688.3 | NP_055503.1 | 4 - 5 | 587 | 94 | |
XM_006717542.3 | XP_006717605.1 | 5 - 6 | 413 | 94 | |
XM_011519762.2 | XP_011518064.1 | 6 - 7 | 746 | 94 | |
XM_017016971.1 | XP_016872460.1 | 5 - 6 | 420 | 94 | |
XM_017016972.1 | XP_016872461.1 | 4 - 5 | 379 | 94 | |
XM_017016973.1 | XP_016872462.1 | 3 - 4 | 3583 | 94 | |
GenBank mRNA | AB449916.1 | - | 2 - 3 | 157 | 94 |
AK292944.1 | - | 4 - 5 | 377 | 94 | |
BC042943.2 | - | 4 - 5 | 425 | 94 | |
CF121539.1 | - | 3 - 4 | 263 | 94 | |
D13644.1 | - | 4 - 5 | 436 | 94 |