The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173620.2 | NP_775891.2 | 2 - 3 | 502 | 56 |
XM_005256347.4 | XP_005256404.1 | 2 - 3 | 226 | 56 | |
XM_017024484.1 | XP_016879973.1 | 2 - 3 | 226 | 56 | |
GenBank mRNA | AK074405.1 | - | 1 - 2 | 65 | 56 |
BC018205.1 | - | 1 - 2 | 97 | 56 | |
BC035561.1 | - | 2 - 3 | 183 | 56 | |
DN999062.1 | - | 2 - 3 | 508 | 56 | |
FM204887.1 | - | 1 - 2 | 86 | 56 |