The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198450.5 | NP_940852.3 | 3 - 4 | 260 | 135 |
XM_011530859.2 | XP_011529161.1 | 3 - 4 | 363 | 135 | |
XM_017029271.1 | XP_016884760.1 | 4 - 5 | 365 | 135 | |
XM_017029272.1 | XP_016884761.1 | 3 - 4 | 300 | 135 | |
XM_017029273.1 | XP_016884762.1 | 3 - 4 | 248 | 135 | |
GenBank mRNA | AK130506.1 | - | 3 - 4 | 326 | 135 |
AY358193.1 | - | 3 - 4 | 253 | 135 | |
BC107096.1 | - | 3 - 4 | 253 | 135 | |
CR749250.1 | - | 3 - 4 | 253 | 135 |