The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198450.5 | NP_940852.3 | 5 - 6 | 409 | 70 |
XM_011530859.2 | XP_011529161.1 | 5 - 6 | 512 | 70 | |
XM_017029271.1 | XP_016884760.1 | 6 - 7 | 514 | 70 | |
XM_017029272.1 | XP_016884761.1 | 5 - 6 | 449 | 70 | |
XM_017029273.1 | XP_016884762.1 | 5 - 6 | 397 | 70 | |
GenBank mRNA | AK130506.1 | - | 5 - 6 | 475 | 70 |
AY358193.1 | - | 5 - 6 | 402 | 70 | |
BC107096.1 | - | 5 - 6 | 402 | 70 | |
CR749250.1 | - | 5 - 6 | 402 | 70 |