The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136195.1 | NP_001129667.1 | 17 - 18 | 2223 | 74 |
NM_001136196.1 | NP_001129668.1 | 16 - 17 | 2050 | 74 | |
NM_013433.4 | NP_038461.2 | 16 - 17 | 2036 | 74 | |
GenBank mRNA | AB210011.1 | - | 15 - 16 | 2603 | 74 |
AF007748.1 | - | 17 - 18 | 2156 | 74 | |
AK056183.1 | - | 16 - 17 | 2562 | 74 | |
AK299485.1 | - | 16 - 17 | 1942 | 74 | |
AK307632.1 | - | 1 - 2 | 63 | 74 | |
BC018314.1 | - | 16 - 17 | 1927 | 74 | |
BC072420.1 | - | 16 - 17 | 2009 | 74 |