The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136195.1 | NP_001129667.1 | 19 - 20 | 2471 | 68 |
NM_001136196.1 | NP_001129668.1 | 18 - 19 | 2298 | 68 | |
NM_013433.4 | NP_038461.2 | 18 - 19 | 2284 | 68 | |
GenBank mRNA | AB210011.1 | - | 17 - 18 | 2851 | 68 |
AF019039.1 | - | 18 - 19 | 2234 | 68 | |
AK299485.1 | - | 18 - 19 | 2190 | 68 | |
AK307632.1 | - | 3 - 4 | 311 | 68 | |
BC018314.1 | - | 18 - 19 | 2175 | 68 | |
BC072420.1 | - | 18 - 19 | 2257 | 68 |